missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidino-1-cyclopentene 97.0+%, TCI America™

$89.48 - $270.29

Chemical Identifiers

CAS 7148-07-4
Molecular Formula C9H15N
Molecular Weight (g/mol) 137.23
MDL Number MFCD00003162
InChI Key KOFSFYBXUYHNJL-UHFFFAOYSA-N
Synonym 1-(1-Cyclopenten-1-yl)pyrrolidine
PubChem CID 81570
IUPAC Name 1-(cyclopenten-1-yl)pyrrolidine
SMILES C1CCN(C1)C2=CCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
P10015G
View Documents
TCI America
P10015G
5 g
Each for $89.48
Only null left
 
P100125G
View Documents
TCI America
P100125G
25 g
Each for $270.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7148-07-4
137.23
KOFSFYBXUYHNJL-UHFFFAOYSA-N
81570
C1CCN(C1)C2=CCCC2
C9H15N
MFCD00003162
1-(1-Cyclopenten-1-yl)pyrrolidine
1-(cyclopenten-1-yl)pyrrolidine

Specifications

7148-07-4
93°C
MFCD00003162
1993
KOFSFYBXUYHNJL-UHFFFAOYSA-N
1-(cyclopenten-1-yl)pyrrolidine
81570
≥97.0% (GC)
1-Pyrrolidino-1-cyclopentene
Yellow
C9H15N
5 g
1-(1-Cyclopenten-1-yl)pyrrolidine
C1CCN(C1)C2=CCCC2
137.23
137.23
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents