missing translation for 'onlineSavingsMsg'
Learn More

1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 946409-40-1
Molecular Formula C12H18Cl2N2O2
Molecular Weight (g/mol) 293.188
MDL Number MFCD06801260
InChI Key FLINLLCJYJQIIY-UHFFFAOYSA-N
Synonym 1-pyridin-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyrid-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid-hydrogen chloride 1/2Show MoreShow Less
PubChem CID 42935493
IUPAC Name 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid;dihydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CN=CC=C2.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69851DA
SDS View Documents
Thermo Scientific Chemicals
CC69851DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69851DE
SDS View Documents
Thermo Scientific Chemicals
CC69851DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'