accessibility menu, dialog, popup

UserName

NOHONHClHCl

CAS RN 946409-40-1

IUPAC Name: 1-[(pyridin-3-yl)methyl]piperidine-4-carboxylic acid dihydrochloride
Synonyms: 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride
Molecular Weight (g/mol): 293.19
Molecular Formula: C12H18Cl2N2O2
InChi Key: FLINLLCJYJQIIY-UHFFFAOYSA-N
SMILES: Cl.Cl.OC(=O)C1CCN(CC2=CN=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)