missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrenemethanol 98.0+%, TCI America™
$82.02 - $229.29
Chemical Identifiers
| CAS | 24463-15-8 |
|---|---|
| Molecular Formula | C17H12O |
| Molecular Weight (g/mol) | 232.282 |
| MDL Number | MFCD00029252 |
| InChI Key | NGDMLQSGYUCLDC-UHFFFAOYSA-N |
| Synonym | 1-(Hydroxymethyl)pyrene |
| PubChem CID | 104977 |
| IUPAC Name | pyren-1-ylmethanol |
| SMILES | C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)CO |
Chemical Identifiers
| 24463-15-8 | |
| 232.282 | |
| NGDMLQSGYUCLDC-UHFFFAOYSA-N | |
| 104977 | |
| C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)CO |
| C17H12O | |
| MFCD00029252 | |
| 1-(Hydroxymethyl)pyrene | |
| pyren-1-ylmethanol |
Specifications
| 24463-15-8 | |
| Yellow | |
| MFCD00029252 | |
| 1-(Hydroxymethyl)pyrene | |
| C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)CO | |
| 232.282 | |
| 232.28 | |
| Crystalline Powder |
| 125°C | |
| C17H12O | |
| 1 g | |
| NGDMLQSGYUCLDC-UHFFFAOYSA-N | |
| pyren-1-ylmethanol | |
| 104977 | |
| ≥98.0% (GC) | |
| 1-Pyrenemethanol |
Safety and Handling
RTECSNumber : UR2457242
TSCA : No