missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenemethanol 98.0+%, TCI America™

Catalog No. P12211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12211G 1 g
P12215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12211G Supplier TCI America Supplier No. P12211G
Only null left

Chemical Identifiers

CAS 24463-15-8
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.282
MDL Number MFCD00029252
InChI Key NGDMLQSGYUCLDC-UHFFFAOYSA-N
Synonym 1-(Hydroxymethyl)pyrene
PubChem CID 104977
IUPAC Name pyren-1-ylmethanol
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)CO

Specifications

Melting Point 125°C
Color Yellow
Quantity 1 g
Formula Weight 232.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Pyrenemethanol
RTECSNumber UR2457242
TSCA No