missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxaldehyde 98.0+%, TCI America™

Catalog No. P14481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14481G 1 g
P14485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14481G Supplier TCI America Supplier No. P14481G

Chemical Identifiers

CAS 3029-19-4
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
MDL Number MFCD00004139
InChI Key RCYFOPUXRMOLQM-UHFFFAOYSA-N
Synonym 1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
PubChem CID 232848
IUPAC Name pyrene-1-carbaldehyde
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O

Specifications

Melting Point 128°C
Quantity 1 g
Formula Weight 230.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Pyrenecarboxaldehyde
RTECSNumber UR2450200
TSCA No