missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxaldehyde, 98+%

Catalog No. AAL1083506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1083506 5 g
AAL1083514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1083506 Supplier Thermo Scientific Chemicals Supplier No. L1083506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3029-19-4 | C17H10O | 230.266 g/mol

1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 3029-19-4
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
MDL Number MFCD00004139
InChI Key RCYFOPUXRMOLQM-UHFFFAOYSA-N
Synonym 1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
PubChem CID 232848
IUPAC Name pyrene-1-carbaldehyde
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Beilstein 1874889
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 1-Pyrenecarboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-196-6
RTECSNumber UR2450200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only