missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenebutyric Acid 98.0+%, TCI America™

Catalog No. p-7135011
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12131G 1 g
P12135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12131G Supplier TCI America Supplier No. P12131G
Only null left

Chemical Identifiers

CAS 3443-45-6
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004141
InChI Key QXYRRCOJHNZVDJ-UHFFFAOYSA-N
Synonym 1-pyrenebutyric acid, 4-pyren-1-yl butanoic acid, 1-pyrenebutanoic acid, 1-pyrenylbutyric acid, pyrene-3-butyric acid, pyrenebutanoic acid, pyrene-1-butyric acid, pyrenebutyric acid, 4-1-pyrenyl butyric acid, gamma-3-pyrenyl butyric acid
PubChem CID 76977
IUPAC Name 4-(pyren-1-yl)butanoic acid
SMILES OC(=O)CCCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 189°C
Color Yellow
Quantity 1 g
Formula Weight 288.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Pyrenebutyric Acid