missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenebutyric acid, 97+%

Catalog No. AC418510050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418510050 5 g Glass bottle
AC418510010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418510050 Supplier Thermo Scientific Chemicals Supplier No. 418510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3443-45-6 | C20H16O2 | 288.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3443-45-6
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004141
InChI Key QXYRRCOJHNZVDJ-UHFFFAOYSA-N
Synonym 1-pyrenebutyric acid, 4-pyren-1-yl butanoic acid, 1-pyrenebutanoic acid, 1-pyrenylbutyric acid, pyrene-3-butyric acid, pyrenebutanoic acid, pyrene-1-butyric acid, pyrenebutyric acid, 4-1-pyrenyl butyric acid, gamma-3-pyrenyl butyric acid
PubChem CID 76977
IUPAC Name 4-pyren-1-ylbutanoic acid
SMILES OC(=O)CCCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 185.0°C to 190.0°C
Color Beige-Yellow or Yellow
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: negligible
Formula Weight 288.34
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material 1-Pyrenebutyric acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-354-7
TSCA TSCA

RUO – Research Use Only