missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenebutyric acid, 97%

Catalog No. AAA1776003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1776003 1 g
AAA1776006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1776003 Supplier Thermo Scientific Chemicals Supplier No. A1776003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3443-45-6 | C20H16O2 | 288.35 g/mol

1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

Solubility
Partially soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3443-45-6
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004141
InChI Key QXYRRCOJHNZVDJ-UHFFFAOYSA-N
Synonym 1-pyrenebutyric acid, 4-pyren-1-yl butanoic acid, 1-pyrenebutanoic acid, 1-pyrenylbutyric acid, pyrene-3-butyric acid, pyrenebutanoic acid, pyrene-1-butyric acid, pyrenebutyric acid, 4-1-pyrenyl butyric acid, gamma-3-pyrenyl butyric acid
PubChem CID 76977
SMILES OC(=O)CCCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 184°C to 186°C
Quantity 1 g
Beilstein 2140554
Solubility Information Partially soluble in water.
IUPAC Name 4-(pyren-1-yl)butanoic acid
Formula Weight 288.35
Percent Purity 97%
Chemical Name or Material 1-Pyrenebutyric acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-354-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only