missing translation for 'onlineSavingsMsg'
Learn More

1-Propynyltri-n-butyltin, 96%

CAS: 64099-82-7 | C15H30Sn | 329.115 g/mol

$112.21 - $411.26

Chemical Identifiers

CAS 64099-82-7
Molecular Formula C15H30Sn
Molecular Weight (g/mol) 329.115
MDL Number MFCD01863650
InChI Key KCQJLTOSSVXOCC-UHFFFAOYSA-N
Synonym tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3
PubChem CID 2733165
IUPAC Name tributyl(prop-1-ynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2020103
View Documents
Thermo Scientific Chemicals
L2020103
1 g N/A
Only null left
 
AAL2020106
View Documents
Thermo Scientific Chemicals
L2020106
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64099-82-7
329.115
KCQJLTOSSVXOCC-UHFFFAOYSA-N
2733165
CCCC[Sn](CCCC)(CCCC)C#CC
C15H30Sn
MFCD01863650
tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3
tributyl(prop-1-ynyl)stannane

Specifications

64099-82-7
277°C
C15H30Sn
H3CC≡CSn[(CH2)3CH3]3
1 g
tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3
CCCC[Sn](CCCC)(CCCC)C#CC
329.115
329.12
1-Propynyltri-n-butyltin
1.082
>110°C (230°F)
1.483
MFCD01863650
UN2788
KCQJLTOSSVXOCC-UHFFFAOYSA-N
tributyl(prop-1-ynyl)stannane
2733165
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H372-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only