Learn More
1-Propynyltri-n-butyltin, 96%
CAS: 64099-82-7 | C15H30Sn | 329.115 g/mol
Supplier: Thermo Scientific Chemicals L2020106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Propynyltri-n-butyltin | |
| 1.082 | |
| >110°C (230°F) | |
| 1.483 | |
| MFCD01863650 | |
| UN2788 | |
| KCQJLTOSSVXOCC-UHFFFAOYSA-N | |
| tributyl(prop-1-ynyl)stannane | |
| 2733165 | |
| 96% |
| 64099-82-7 | |
| 277°C | |
| C15H30Sn | |
| H3CC≡CSn[(CH2)3CH3]3 | |
| 5 g | |
| tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3 | |
| CCCC[Sn](CCCC)(CCCC)C#CC | |
| 329.115 | |
| 329.12 |
Chemical Identifiers
| 64099-82-7 | |
| 329.115 | |
| KCQJLTOSSVXOCC-UHFFFAOYSA-N | |
| 2733165 | |
| CCCC[Sn](CCCC)(CCCC)C#CC |
| C15H30Sn | |
| MFCD01863650 | |
| tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3 | |
| tributyl(prop-1-ynyl)stannane |
Safety and Handling
GHS H Statement
H300-H311-H372-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only