missing translation for 'onlineSavingsMsg'
Learn More

1-Propylpiperazine Dihydrobromide 98.0+%, TCI America™

Catalog No. P164525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P164525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P164525G Supplier TCI America Supplier No. P164525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64262-23-3
Molecular Formula C7H18Br2N2
Molecular Weight (g/mol) 290.043
InChI Key YTPQLWVHCBATKO-UHFFFAOYSA-N
Synonym 1-propylpiperazine dihydrobromide, 1-n-propylpiperazine dihydrobromide, n-propylpiperazinium dibromide, n-propylpiperazine dihydrobromide, 1-n-propylpiperazine 2hbr, piperazine, 1-propyl-, dihydrobromide, acmc-209nl5, ksc494c5l, propylpiperazine, bromide, bromide, 1-n-propyl-piperazine dihydrobromide
PubChem CID 3085288
IUPAC Name 1-propylpiperazine;dihydrobromide
SMILES CCCN1CCNCC1.Br.Br

Specifications

Melting Point 254°C
Color White
Quantity 25 g
Formula Weight 290.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Propylpiperazine Dihydrobromide