accessibility menu, dialog, popup

UserName

CAS RN 64262-23-3

IUPAC Name: dihydrogen 1-propylpiperazine dibromide
Synonyms: dihydrogen 1-propylpiperazine dibromide
Molecular Weight (g/mol): 290.04
Molecular Formula: C7H18Br2N2
InChi Key: YTPQLWVHCBATKO-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].CCCN1CCNCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results