missing translation for 'onlineSavingsMsg'
Learn More

1-Propylamine, 98%

CAS: 107-10-8 | C3H9N | 59.112 g/mol

Supplier:  Thermo Scientific Chemicals A116060F

 View more versions of this product

Catalog No. AAA116060F


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.

Solubility
Miscible with water, alcohol, ether

Notes
Miscible with water, alcohol, ether
Specifications

Specifications

1-Propylamine
-83°C
12.6
−12°C (10°F)
C3H9N
MFCD00008205
UN1277
Air Sensitive
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propan-1-amine
7852
59.11
107-10-8
0.719
48°C to 49°C
Amine-like
1.389
2500 mL
1098243
14,7843
Miscible with water,alcohol,ether
CCCN
59.112
CHEBI:39870
98%

Chemical Identifiers

107-10-8
59.112
WGYKZJWCGVVSQN-UHFFFAOYSA-N
7852
propan-1-amine
C3H9N
MFCD00008205
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
CHEBI:39870
CCCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H314-H318-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c

H225-H290-H302-H311+H331-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYLAMINE

EINECSNumber : 203-462-3

RTECSNumber : UH9100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only