missing translation for 'onlineSavingsMsg'
Learn More

1-Propylamine, 98%

Catalog No. p-7081576
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11606AP 500 mL
AAA116060F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11606AP Supplier Thermo Scientific Chemicals Supplier No. A11606AP

CAS: 107-10-8 | C3H9N | 59.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.

Solubility
Miscible with water, alcohol, ether

Notes
Miscible with water, alcohol, ether

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN

Specifications

Melting Point -83°C
Density 0.719
pH 12.6
Boiling Point 48°C to 49°C
Flash Point −12°C (10°F)
Odor Amine-like
Refractive Index 1.389
Quantity 500 mL
UN Number UN1277
Beilstein 1098243
Sensitivity Air Sensitive
Merck Index 14,7843
Solubility Information Miscible with water,alcohol,ether
Formula Weight 59.11
Percent Purity 98%
Chemical Name or Material 1-Propylamine
Show More Show Less
Hazard Category H225-H290-H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H225-H311-H314-H318-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYLAMINE
EINECSNumber 203-462-3
RTECSNumber UH9100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only