missing translation for 'onlineSavingsMsg'
Learn More

1-Propyl isothiocyanate, 97%

Catalog No. AAA1449906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449906 5 g
AAA1449914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1449906 Supplier Thermo Scientific Chemicals Supplier No. A1449906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 628-30-8 | C4H7NS | 101.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-30-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00004823
InChI Key KKASGUHLXWAKEZ-UHFFFAOYSA-N
Synonym propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
PubChem CID 69403
IUPAC Name 1-isothiocyanatopropane
SMILES CCCN=C=S

Specifications

Density 0.977
Boiling Point 152°C to 154°C
Flash Point 46°C (114°F)
Odor Pungent
Refractive Index 1.504
Quantity 5 g
UN Number UN2924
Beilstein 1740199
Sensitivity Moisture sensitive
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material 1-Propyl isothiocyanate
Show More Show Less
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 211-035-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only