missing translation for 'onlineSavingsMsg'
Learn More

1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. P22771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22771G 1 g
P22775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22771G Supplier TCI America Supplier No. P22771G
Only null left

Chemical Identifiers

CAS 827614-69-7
Molecular Formula C12H21BN2O2
Molecular Weight (g/mol) 236.12
MDL Number MFCD05663871
InChI Key BKLGYJWLZWMIDO-UHFFFAOYSA-N
Synonym 1-propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-propyl-1h-pyrazole-4-boronic acid pinacol ester, 1-propyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-propylpyrazole-4-boronic acid pinacol ester, 1h-pyrazole, 1-propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-propyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-propyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-propyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-propyl-1h-pyrazole
PubChem CID 2760487
IUPAC Name 1-propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CCCN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 1 g
Formula Weight 236.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole