accessibility menu, dialog, popup

UserName

CH3NNBOCH3CH3CH3H3CO

CAS RN 827614-69-7

IUPAC Name: 1-propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 236.12
Molecular Formula: C12H21BN2O2
InChi Key: BKLGYJWLZWMIDO-UHFFFAOYSA-N
SMILES: CCCN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results