missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propene 1,3-Sultone 99.0+%, TCI America™
$152.08 - $508.25
Chemical Identifiers
| CAS | 21806-61-1 |
|---|---|
| Molecular Formula | C3H4O3S |
| Molecular Weight (g/mol) | 120.122 |
| MDL Number | MFCD12405143 |
| InChI Key | KLLQVNFCMHPYGL-UHFFFAOYSA-N |
| Synonym | 5H-1,2-Oxathiole 2,2-Dioxide |
| PubChem CID | 10898703 |
| IUPAC Name | 5H-oxathiole 2,2-dioxide |
| SMILES | C1C=CS(=O)(=O)O1 |
Chemical Identifiers
| 21806-61-1 | |
| 120.122 | |
| KLLQVNFCMHPYGL-UHFFFAOYSA-N | |
| 10898703 | |
| C1C=CS(=O)(=O)O1 |
| C3H4O3S | |
| MFCD12405143 | |
| 5H-1,2-Oxathiole 2,2-Dioxide | |
| 5H-oxathiole 2,2-dioxide |
Specifications
| 21806-61-1 | |
| White | |
| C3H4O3S | |
| 5 g | |
| KLLQVNFCMHPYGL-UHFFFAOYSA-N | |
| 5H-oxathiole 2,2-dioxide | |
| 10898703 | |
| ≥99.0% (GC) | |
| 1-Propene 1,3-Sultone |
| 85°C | |
| 118°C | |
| MFCD12405143 | |
| 5H-1,2-Oxathiole 2,2-Dioxide | |
| C1C=CS(=O)(=O)O1 | |
| 120.122 | |
| 120.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No