accessibility menu, dialog, popup

UserName

CAS RN 21806-61-1

IUPAC Name: 5H-1,2λ⁶-oxathiole-2,2-dione
Synonyms: 5H-1,2λ⁶-oxathiole-2,2-dione
Molecular Weight (g/mol): 120.12
Molecular Formula: C3H4O3S
InChi Key: KLLQVNFCMHPYGL-UHFFFAOYSA-N
SMILES: O=S1(=O)OCC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results