missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018788
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018788 5 mL
111018349 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018788 Supplier Merck Emd Millipore Supplier No. 965665MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 71-23-8
Vapor Pressure 10 mmHg (147°C)
Vapor Density 2.1 (vs. air)
Density 0.804 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH2OH
MDL Number MFCD00002941
Melting Point -127°C
Boiling Point 97°C
Synonym Propyl alcohol
Quantity 5 mL
Formula Weight 60.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less