missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 80360-23-2 |
---|---|
Molecular Formula | C15H11NO3S |
Molecular Weight (g/mol) | 285.32 |
MDL Number | MFCD03086091 |
InChI Key | NWVOBVXBWUGTTO-UHFFFAOYSA-N |
Synonym | 1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde |
PubChem CID | 2776223 |
IUPAC Name | 1-(benzenesulfonyl)indole-2-carbaldehyde |
SMILES | O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC03304CB
|
Thermo Scientific Chemicals
CC03304CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC03304DA
|
Thermo Scientific Chemicals
CC03304DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
80360-23-2 | |
285.32 | |
NWVOBVXBWUGTTO-UHFFFAOYSA-N | |
2776223 | |
O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1 |
C15H11NO3S | |
MFCD03086091 | |
1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde | |
1-(benzenesulfonyl)indole-2-carbaldehyde |
Specifications
80360-23-2 | |
100.0 | |
C15H11NO3S | |
250mg | |
NWVOBVXBWUGTTO-UHFFFAOYSA-N | |
1-(benzenesulfonyl)indole-2-carbaldehyde | |
2776223 | |
97% |
96.5 | |
97% | |
MFCD03086091 | |
1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde | |
O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1 | |
285.32 | |
285.32 | |
1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde |