missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC03304CB
Specifications
| 1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde | |
| 96.5 | |
| 97% | |
| MFCD03086091 | |
| 1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde | |
| O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1 | |
| 285.32 | |
| 285.32 |
| 80360-23-2 | |
| 100.0 | |
| C15H11NO3S | |
| 250mg | |
| NWVOBVXBWUGTTO-UHFFFAOYSA-N | |
| 1-(benzenesulfonyl)indole-2-carbaldehyde | |
| 2776223 | |
| 97% |
Chemical Identifiers
| 80360-23-2 | |
| 285.32 | |
| NWVOBVXBWUGTTO-UHFFFAOYSA-N | |
| 2776223 | |
| O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1 |
| C15H11NO3S | |
| MFCD03086091 | |
| 1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde | |
| 1-(benzenesulfonyl)indole-2-carbaldehyde |