Learn More
1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid, 97%, May contain varying amounts of anhydri, Thermo Scientific™
$163.66 - $163.66
Chemical Identifiers
| CAS | 129271-98-3 |
|---|---|
| Molecular Formula | C14H12BNO4S |
| Molecular Weight (g/mol) | 301.12 |
| MDL Number | MFCD02681892 |
| InChI Key | YKTZLHLBQGCFQX-UHFFFAOYSA-N |
| Synonym | 1-phenylsulfonyl-3-indoleboronic acid, 1-phenylsulfonyl-1h-indol-3-ylboronic acid, 1-phenylsulfonyl-1h-indol-3-yl boronic acid, 1-benzenesulfonyl indol-3-yl boronic acid, 1-phenylsulfonylindole-3-boronic acid, 1-phenylsulphonyl-1h-indole-3-boronic acid, 1-phenylsulfonyl-3-indolylboronic acid, boronic acid, b-1-phenylsulfonyl-1h-indol-3-yl, 1-benzenesulfonyl indol-3-ylboronic acid, 1-benzenesulfonyl-1h-indol-3-yl boranediol |
| PubChem CID | 2776217 |
| IUPAC Name | [1-(benzenesulfonyl)indol-3-yl]boronic acid |
| SMILES | OB(O)C1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 129271-98-3 | |
| 301.12 | |
| YKTZLHLBQGCFQX-UHFFFAOYSA-N | |
| 2776217 | |
| OB(O)C1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 |
| C14H12BNO4S | |
| MFCD02681892 | |
| 1-phenylsulfonyl-3-indoleboronic acid, 1-phenylsulfonyl-1h-indol-3-ylboronic acid, 1-phenylsulfonyl-1h-indol-3-yl boronic acid, 1-benzenesulfonyl indol-3-yl boronic acid, 1-phenylsulfonylindole-3-boronic acid, 1-phenylsulphonyl-1h-indole-3-boronic acid, 1-phenylsulfonyl-3-indolylboronic acid, boronic acid, b-1-phenylsulfonyl-1h-indol-3-yl, 1-benzenesulfonyl indol-3-ylboronic acid, 1-benzenesulfonyl-1h-indol-3-yl boranediol | |
| [1-(benzenesulfonyl)indol-3-yl]boronic acid |
Specifications
| 129271-98-3 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02681892 | |
| 1-phenylsulfonyl-3-indoleboronic acid, 1-phenylsulfonyl-1h-indol-3-ylboronic acid, 1-phenylsulfonyl-1h-indol-3-yl boronic acid, 1-benzenesulfonyl indol-3-yl boronic acid, 1-phenylsulfonylindole-3-boronic acid, 1-phenylsulphonyl-1h-indole-3-boronic acid, 1-phenylsulfonyl-3-indolylboronic acid, boronic acid, b-1-phenylsulfonyl-1h-indol-3-yl, 1-benzenesulfonyl indol-3-ylboronic acid, 1-benzenesulfonyl-1h-indol-3-yl boranediol | |
| OB(O)C1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 | |
| 301.12 | |
| 301.13 | |
| 1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid, May contain varying amounts of anhydride |
| 96.5 | |
| 97% | |
| C14H12BNO4S | |
| 1 g | |
| YKTZLHLBQGCFQX-UHFFFAOYSA-N | |
| [1-(benzenesulfonyl)indol-3-yl]boronic acid | |
| 2776217 | |
| 97% |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning