missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC03312DE
Specifications
| 1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid | |
| 97% | |
| C14H12BNO4S | |
| 1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid | |
| B(C1=CC2=CC=CC=C2N1S(=O)(=O)C3=CC=CC=C3)(O)O | |
| 301.123 | |
| 301.13 |
| 342404-46-0 | |
| Amber Glass Bottle | |
| 5g | |
| HXWLCYMHOULBJZ-UHFFFAOYSA-N | |
| [1-(benzenesulfonyl)indol-2-yl]boronic acid | |
| 2776228 |
Chemical Identifiers
| 342404-46-0 | |
| 301.123 | |
| 1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid | |
| [1-(benzenesulfonyl)indol-2-yl]boronic acid |
| C14H12BNO4S | |
| HXWLCYMHOULBJZ-UHFFFAOYSA-N | |
| 2776228 | |
| B(C1=CC2=CC=CC=C2N1S(=O)(=O)C3=CC=CC=C3)(O)O |