missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylsemicarbazide, 99%

Catalog No. p-7026367
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2233114 25 g
AAB2233122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2233114 Supplier Thermo Scientific Chemicals Supplier No. B2233114
Only null left

CAS: 103-03-7 | C7H9N3O | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-03-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007942
InChI Key AVKHCKXGKPAGEI-UHFFFAOYSA-N
Synonym phenicarbazide, 1-phenylsemicarbazide, 2-phenylhydrazinecarboxamide, phenicarbazid, phenylsemicarbazide, carbaphen, kryogenin, phenygenine, febrimin, 1-carbamyl-2-phenylhydrazine
PubChem CID 61002
ChEBI CHEBI:82521
IUPAC Name anilinourea
SMILES NC(=O)NNC1=CC=CC=C1

Specifications

Melting Point 171°C to 173°C
Odor Odorless
Quantity 25 g
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 1-Phenylsemicarbazide
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 203-072-3
RTECSNumber FD0575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only