missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrazole 98.0+%, TCI America™

Supplier:  TCI America P186825G

 View more versions of this product

Catalog No. P186825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Phenylpyrazole
11°C
142°C
MFCD00003112
1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole
C1=CN(N=C1)C1=CC=CC=C1
144.18
CHEBI:38879
≥98.0% (GC)
1126-00-7
Yellow
C9H8N2
25 g
WITMXBRCQWOZPX-UHFFFAOYSA-N
1-phenyl-1H-pyrazole
70769
144.18
Liquid

Chemical Identifiers

1126-00-7
144.18
WITMXBRCQWOZPX-UHFFFAOYSA-N
70769
1-phenyl-1H-pyrazole
C9H8N2
MFCD00003112
1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole
CHEBI:38879
C1=CN(N=C1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions