missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrazole 98.0+%, TCI America™

Catalog No. p-7135297
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18685G 5 g
P186825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18685G Supplier TCI America Supplier No. P18685G
Only null left

Chemical Identifiers

CAS 1126-00-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00003112
InChI Key WITMXBRCQWOZPX-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole
PubChem CID 70769
ChEBI CHEBI:38879
IUPAC Name 1-phenyl-1H-pyrazole
SMILES C1=CN(N=C1)C1=CC=CC=C1

Specifications

Melting Point 11°C
Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenylpyrazole