missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyloxindole 98.0+%, TCI America™

Catalog No. P17695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17695G 5 g
P176925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17695G Supplier TCI America Supplier No. P17695G

Chemical Identifiers

CAS 3335-98-6
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.25
MDL Number MFCD00234850
InChI Key OWPNVXATCSXTBK-UHFFFAOYSA-N
Synonym 1-Phenyl-2-indolinone, 2-Oxo-1-phenylindoline
PubChem CID 314787
IUPAC Name 1-phenyl-2,3-dihydro-1H-indol-2-one
SMILES O=C1CC2=CC=CC=C2N1C1=CC=CC=C1

Specifications

Melting Point 122°C
Color Yellow
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyloxindole