accessibility menu, dialog, popup

UserName

ON

CAS RN 3335-98-6

IUPAC Name: 1-phenyl-2,3-dihydro-1H-indol-2-one
Synonyms: 1-phenyl-3H-indol-2-one
Molecular Weight (g/mol): 209.25
Molecular Formula: C14H11NO
InChi Key: OWPNVXATCSXTBK-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results