missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenylisatin 98.0+%, TCI America™
Supplier: TCI America P227925G
Specifications
| 1-Phenylisatin | |
| 141°C | |
| MFCD00082681 | |
| 1-phenylisatin, 1-phenylindoline-2,3-dione, 1-phenyl-1h-indole-2,3-dione, 1h-indole-2,3-dione, 1-phenyl, 1-phenyl-indole-2,3-dione, indole-2,3-dione, 1-phenyl, 1h-indole-2,3-dione, 1-phenyl-9ci, 1-phenyl-2,3-dihydro-1h-indole-2,3-dione, phenylisatin, n-phenylisatin | |
| C1=CC=C(C=C1)N2C3=CC=CC=C3C(=O)C2=O | |
| 223.231 | |
| 223.23 | |
| Crystalline Powder |
| 723-89-7 | |
| C14H9NO2 | |
| 25 g | |
| UWCPWBIMRYXUOU-UHFFFAOYSA-N | |
| 1-phenylindole-2,3-dione | |
| 12884 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 723-89-7 | |
| 223.231 | |
| UWCPWBIMRYXUOU-UHFFFAOYSA-N | |
| 12884 | |
| C1=CC=C(C=C1)N2C3=CC=CC=C3C(=O)C2=O |
| C14H9NO2 | |
| MFCD00082681 | |
| 1-phenylisatin, 1-phenylindoline-2,3-dione, 1-phenyl-1h-indole-2,3-dione, 1h-indole-2,3-dione, 1-phenyl, 1-phenyl-indole-2,3-dione, indole-2,3-dione, 1-phenyl, 1h-indole-2,3-dione, 1-phenyl-9ci, 1-phenyl-2,3-dihydro-1h-indole-2,3-dione, phenylisatin, n-phenylisatin | |
| 1-phenylindole-2,3-dione |
Safety and Handling
RTECSNumber : NL7984000
TSCA : No