missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylisatin 98.0+%, TCI America™

Catalog No. p-7135490
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22795G 5 g
P227925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22795G Supplier TCI America Supplier No. P22795G

Chemical Identifiers

CAS 723-89-7
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00082681
InChI Key UWCPWBIMRYXUOU-UHFFFAOYSA-N
Synonym 1-phenylisatin, 1-phenylindoline-2,3-dione, 1-phenyl-1h-indole-2,3-dione, 1h-indole-2,3-dione, 1-phenyl, 1-phenyl-indole-2,3-dione, indole-2,3-dione, 1-phenyl, 1h-indole-2,3-dione, 1-phenyl-9ci, 1-phenyl-2,3-dihydro-1h-indole-2,3-dione, phenylisatin, n-phenylisatin
PubChem CID 12884
IUPAC Name 1-phenylindole-2,3-dione
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3C(=O)C2=O

Specifications

Melting Point 141°C
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenylisatin
RTECSNumber NL7984000
TSCA No