missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarbonitrile 98.0+%, TCI America™

Catalog No. p-7135427
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21461G 1 g
P21465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21461G Supplier TCI America Supplier No. P21461G

Chemical Identifiers

CAS 77-57-6
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00001358
InChI Key GDXMFFGTPGAGGX-UHFFFAOYSA-N
Synonym 1-phenylcyclopentanecarbonitrile, 1-phenyl-1-cyclopentanecarbonitrile, 1-phenyl-cyclopentanecarbonitrile, cyclopentanecarbonitrile, 1-phenyl, 1-phenylcyclopentanenitrile, phenylcyclopentane carbonitrile, gdxmffgtpgaggx-uhfffaoysa
PubChem CID 66168
IUPAC Name 1-phenylcyclopentane-1-carbonitrile
SMILES C1CCC(C1)(C#N)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenylcyclopentanecarbonitrile
EINECSNumber (9)-1174
TSCA No