missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenylcyclobutanecarbonitrile 98.0+%, TCI America™
Supplier: TCI America P225225G
Specifications
| 1-Phenylcyclobutanecarbonitrile | |
| Yellow | |
| C11H11N | |
| 25 g | |
| DHIDUDPFTZJPCQ-UHFFFAOYSA-N | |
| 1-phenylcyclobutane-1-carbonitrile | |
| 84400 | |
| ≥98.0% (GC) |
| 14377-68-5 | |
| 85°C | |
| MFCD00019258 | |
| 1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile | |
| C1CC(C1)(C#N)C2=CC=CC=C2 | |
| 157.216 | |
| 157.22 | |
| Liquid |
Chemical Identifiers
| 14377-68-5 | |
| 157.216 | |
| DHIDUDPFTZJPCQ-UHFFFAOYSA-N | |
| 1-phenylcyclobutane-1-carbonitrile |
| C11H11N | |
| MFCD00019258 | |
| 84400 | |
| C1CC(C1)(C#N)C2=CC=CC=C2 |
Safety and Handling
TSCA : No