missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenylcyclobutanecarbonitrile 98.0+%, TCI America™
$121.28 - $415.84
Chemical Identifiers
| CAS | 14377-68-5 |
|---|---|
| Molecular Formula | C11H11N |
| Molecular Weight (g/mol) | 157.216 |
| MDL Number | MFCD00019258 |
| InChI Key | DHIDUDPFTZJPCQ-UHFFFAOYSA-N |
| PubChem CID | 84400 |
| IUPAC Name | 1-phenylcyclobutane-1-carbonitrile |
| SMILES | C1CC(C1)(C#N)C2=CC=CC=C2 |
Chemical Identifiers
| 14377-68-5 | |
| 157.216 | |
| DHIDUDPFTZJPCQ-UHFFFAOYSA-N | |
| 1-phenylcyclobutane-1-carbonitrile |
| C11H11N | |
| MFCD00019258 | |
| 84400 | |
| C1CC(C1)(C#N)C2=CC=CC=C2 |
Specifications
| 14377-68-5 | |
| 85°C | |
| MFCD00019258 | |
| 1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile | |
| C1CC(C1)(C#N)C2=CC=CC=C2 | |
| 157.216 | |
| 157.22 | |
| Liquid |
| Yellow | |
| C11H11N | |
| 5 g | |
| DHIDUDPFTZJPCQ-UHFFFAOYSA-N | |
| 1-phenylcyclobutane-1-carbonitrile | |
| 84400 | |
| ≥98.0% (GC) | |
| 1-Phenylcyclobutanecarbonitrile |
Safety and Handling
TSCA : No