missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%

Catalog No. p-7025134
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2047803 1 g
AAB2047806 5 g
AAB2047814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2047803 Supplier Thermo Scientific Chemicals Supplier No. B2047803
Only null left

CAS: 321-07-3 | C10H7F3N2O | 228.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 321-07-3
Molecular Formula C10H7F3N2O
Molecular Weight (g/mol) 228.174
MDL Number MFCD00051655
InChI Key GLGRRRKQSFURGD-UHFFFAOYSA-N
Synonym 1-phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 1-phenyl-3-trifluoromethyl-1h-pyrazol-5 4h-one, 2-phenyl-5-trifluoromethyl-4h-pyrazol-3-one, 2-phenyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one, 1-phenyl-3-trifluoromethyl-4,5-dihydro-1h-pyrazol-5-one, maybridge4_000200, acmc-1ai73, 1-phenyl-3-trifluoromethyl-2-pyrazoline-5-one, 2-phenyl-5-trifluoromethyl-2,4-dihydro-pyrazol-3-one
PubChem CID 668302
IUPAC Name 2-phenyl-5-(trifluoromethyl)-4H-pyrazol-3-one
SMILES C1C(=NN(C1=O)C2=CC=CC=C2)C(F)(F)F

Specifications

Melting Point 191°C to 195°C
Quantity 1 g
Beilstein 655085
Formula Weight 228.18
Percent Purity 98%
Chemical Name or Material 1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only