CAS RN 321-07-3
IUPAC Name:
1-phenyl-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one
Synonyms:
2-phenyl-5-(trifluoromethyl)-4H-pyrazol-3-one
Molecular Weight (g/mol):
228.17
Molecular Formula:
C10H7F3N2O
InChi Key:
GLGRRRKQSFURGD-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NN(C(=O)C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%
1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%
CAS: 321-07-3 Molecular Formula: C10H7F3N2O Molecular Weight (g/mol): 228.174 MDL Number: MFCD00051655 InChI Key: GLGRRRKQSFURGD-UHFFFAOYSA-N Synonym: 1-phenyl-3-trifluoromethyl-2-pyrazolin-5-one,1-phenyl-3-trifluoromethyl-1h-pyrazol-5 4h-one,2-phenyl-5-trifluoromethyl-4h-pyrazol-3-one,2-phenyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one,1-phenyl-3-trifluoromethyl-4,5-dihydro-1h-pyrazol-5-one,maybridge4_000200,acmc-1ai73,1-phenyl-3-trifluoromethyl-2-pyrazoline-5-one,2-phenyl-5-trifluoromethyl-2,4-dihydro-pyrazol-3-one PubChem CID: 668302 IUPAC Name: 2-phenyl-5-(trifluoromethyl)-4H-pyrazol-3-one SMILES: C1C(=NN(C1=O)C2=CC=CC=C2)C(F)(F)F