missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-pyrazolidone 98.0+%, TCI America™

Catalog No. P015325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P015325G 25 g
P0153500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P015325G Supplier TCI America Supplier No. P015325G

Chemical Identifiers

CAS 92-43-3
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00003094
InChI Key CMCWWLVWPDLCRM-UHFFFAOYSA-N
Synonym phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
PubChem CID 7090
IUPAC Name 1-phenylpyrazolidin-3-one
SMILES C1CN(NC1=O)C2=CC=CC=C2

Specifications

Melting Point 122°C
Quantity 25 g
UN Number 2811
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-pyrazolidone
EINECSNumber (9)-1855
RTECSNumber UQ8750000
TSCA Yes