accessibility menu, dialog, popup

UserName

CAS RN 92-43-3

IUPAC Name: 1-phenylpyrazolidin-3-one
Synonyms: phenidone
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: CMCWWLVWPDLCRM-UHFFFAOYSA-N
SMILES: O=C1CCN(N1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)