missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-pyrazolidinone, 97%

Catalog No. p-7081903
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1208930 250 g
AAA120890B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1208930 Supplier Thermo Scientific Chemicals Supplier No. A1208930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-43-3 | C9H10N2O | 162.192 g/mol

Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 92-43-3
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00003094
InChI Key CMCWWLVWPDLCRM-UHFFFAOYSA-N
Synonym phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
PubChem CID 7090
IUPAC Name 1-phenylpyrazolidin-3-one
SMILES C1CN(NC1=O)C2=CC=CC=C2

Specifications

Melting Point 120°C to 123°C
Quantity 250 g
UN Number UN2811
Beilstein 131856
Sensitivity Light sensitive
Merck Index 14,7309
Solubility Information Soluble in water (1:10 hot),methanol (0.1mg/mL),DMSO (75mg/mL),and ethanol.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1-Phenyl-3-pyrazolidinone
Hazard Category H301
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-155-1
RTECSNumber UQ8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only