missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline 98.0+%, TCI America™

Catalog No. p-7135327
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19511G 1 g
P19515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19511G Supplier TCI America Supplier No. P19511G

Chemical Identifiers

CAS 60078-97-9
Molecular Formula C18H20N2
Molecular Weight (g/mol) 264.37
MDL Number MFCD00153039
InChI Key BJEAUUFMQKUXAX-UHFFFAOYSA-N
Synonym 3-Mesityl-1-phenyl-2-pyrazoline
PubChem CID 108884
IUPAC Name 1-phenyl-3-(2,4,6-trimethylphenyl)-4,5-dihydro-1H-pyrazole
SMILES CC1=CC(C)=C(C2=NN(CC2)C2=CC=CC=C2)C(C)=C1

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 264.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline