accessibility menu, dialog, popup

UserName

CH3CH3H3CNN

CAS RN 60078-97-9

IUPAC Name: 1-phenyl-3-(2,4,6-trimethylphenyl)-4,5-dihydro-1H-pyrazole
Synonyms: 1-phenyl-3-(2,4,6-trimethylphenyl)-4,5-dihydropyrazole
Molecular Weight (g/mol): 264.37
Molecular Formula: C18H20N2
InChi Key: BJEAUUFMQKUXAX-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C2=NN(CC2)C2=CC=CC=C2)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results