missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-trimethylsilyloxyethylene 95.0+%, TCI America™

Catalog No. P128010ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P128010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P128010ML Supplier TCI America Supplier No. P128010ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13735-81-4
Molecular Formula C11H16OSi
Molecular Weight (g/mol) 192.333
MDL Number MFCD00008582
InChI Key AFFPCIMDERUIST-UHFFFAOYSA-N
Synonym 1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf
PubChem CID 117406
IUPAC Name trimethyl(1-phenylethenoxy)silane
SMILES C[Si](C)(C)OC(=C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 10 mL
Formula Weight 192.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-trimethylsilyloxyethylene