missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne 97.0+%, TCI America™

Catalog No. p-7134497
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
P02195ML 5 mL
P021925ML 25 mL
2 options

Catalog No. P02195ML

Supplier: TCI America P02195ML

Only null left

Chemical Identifiers

CAS 673-32-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00009272
InChI Key GHUURDQYRGVEHX-UHFFFAOYSA-N
Synonym 1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
PubChem CID 69601
IUPAC Name prop-1-ynylbenzene
SMILES CC#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 181°C
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propyne