missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne, 99%

CAS: 673-32-5 | C9H8 | 116.16 g/mol

$134.89 - $134.89

Chemical Identifiers

CAS 673-32-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009272
InChI Key GHUURDQYRGVEHX-UHFFFAOYSA-N
Synonym 1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
PubChem CID 69601
IUPAC Name prop-1-ynylbenzene
SMILES CC#CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC171320050
SDS View Documents
Thermo Scientific Chemicals
171320050
5 g Glass bottle
Each for $134.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

673-32-5
116.16
GHUURDQYRGVEHX-UHFFFAOYSA-N
69601
CC#CC1=CC=CC=C1
C9H8
MFCD00009272
1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
prop-1-ynylbenzene

Specifications

673-32-5
100.0
0.9200g/mL
62°C
98.5% min. (GC)
C9H8
C6H5C≡CCH3
5 g
0.92
GHUURDQYRGVEHX-UHFFFAOYSA-N
prop-1-ynylbenzene
69601
99%
1-Phenyl-1-propyne
98.5
Yellow
185.0°C
Authentic
Glass bottle
1.5630 to 1.5650
MFCD00009272
05, 514
1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
CC#CC1=CC=CC=C1
116.16
116.16
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-607-7

SDS
Documents

Documents

RUO – Research Use Only