missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-propanol 98.0+%, TCI America™
$50.33 - $356.03
Chemical Identifiers
| CAS | 93-54-9 |
|---|---|
| Molecular Formula | C9H12O |
| Molecular Weight (g/mol) | 136.194 |
| MDL Number | MFCD00004564 |
| InChI Key | DYUQAZSOFZSPHD-UHFFFAOYSA-N |
| Synonym | 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon |
| PubChem CID | 7147 |
| IUPAC Name | 1-phenylpropan-1-ol |
| SMILES | CCC(C1=CC=CC=C1)O |
Chemical Identifiers
| 93-54-9 | |
| 136.194 | |
| DYUQAZSOFZSPHD-UHFFFAOYSA-N | |
| 7147 | |
| CCC(C1=CC=CC=C1)O |
| C9H12O | |
| MFCD00004564 | |
| 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon | |
| 1-phenylpropan-1-ol |
Specifications
| 93-54-9 | |
| 212°C | |
| MFCD00004564 | |
| 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon | |
| CCC(C1=CC=CC=C1)O | |
| 136.194 | |
| 136.19 | |
| Liquid |
| Yellow | |
| C9H12O | |
| 25 g | |
| DYUQAZSOFZSPHD-UHFFFAOYSA-N | |
| 1-phenylpropan-1-ol | |
| 7147 | |
| ≥98.0% (GC) | |
| 1-Phenyl-1-propanol |
Safety and Handling
EINECSNumber : (3)-1034&(3)-1049
RTECSNumber : DO5470000
TSCA : No