missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propanol 98.0+%, TCI America™

Catalog No. P0212500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0212500G 500 g
P021225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0212500G Supplier TCI America Supplier No. P0212500G

Chemical Identifiers

CAS 93-54-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004564
InChI Key DYUQAZSOFZSPHD-UHFFFAOYSA-N
Synonym 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
PubChem CID 7147
IUPAC Name 1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 212°C
Quantity 500 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propanol
EINECSNumber (3)-1034&(3)-1049
RTECSNumber DO5470000
TSCA No