accessibility menu, dialog, popup

UserName

CAS RN 1817-51-2

IUPAC Name: 1-phenylhex-1-yn-3-ol
Synonyms: 1-phenylhex-1-yn-3-ol
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: HGUIQUVFOYTZNC-UHFFFAOYNA-N
SMILES: CCCC(O)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)