missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-hexanol 98.0+%, TCI America™

Catalog No. p-7135205
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16835G 5 g
P168325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16835G Supplier TCI America Supplier No. P16835G

Chemical Identifiers

CAS 4471-05-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD02258728
InChI Key SVCRDVHXRDRHCP-UHFFFAOYNA-N
Synonym alpha-Pentylbenzyl Alcohol
PubChem CID 221292
IUPAC Name 1-phenylhexan-1-ol
SMILES CCCCCC(O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-hexanol